VWR®, Laboratory Jacks
Supplier: VWR Collection
VWRI245-1222EA 415 SGD
VWRI245-1222 VWRI245-1223 VWRI245-1220 VWRI245-1221 VWRI245-1228 VWRI245-1227 VWRI245-1226
VWR®, Laboratory Jacks
Laboratory Jacks
High quality, stable and inexpensive range of lifting platforms for a variety of uses. With plates made from matt, brushed, non rusting stainless steel, and smooth running aluminium frame with scissor action. Rubber feet ensure stable support.
  • Ergonomically-designed height adjustment knob for easy operation
  • Available with several different platform sizes
  • M10 hole for retort rod (except 245-1220)