VWR®, Sample Dippers
Supplier: VWR Collection
VWRI331-3413EA 586 SGD
VWRI331-3413 VWRI331-0293 VWRI331-0292 VWRI331-0294
VWR®, Sample Dippers
Samplers Sample Dippers
All sample dippers are made of blue PP and consist of a bottle with snap handle.
  • Containers designed for sample collection and dispatch for testing in the same container, eliminating risk of cross-contamination
  • Sterilised by electron-beam irradiation and individually packed in sealed zip bag to guarantee integrity of sample
Ordering information: Available with either hinged plug or screw cap.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Order Now
SPECIFICATIONS
Environmentally Preferable