VWR® INCU-Line® IL 10, IL 23, IL 56, Digital Incubators
Supplier: VWR Collection
VWRI390-0384EA 1250 SGD
VWRI390-0384 VWRI390-0385 VWRI390-0482 VWRI390-0999
VWR® INCU-Line® IL 10, IL 23, IL 56, Digital Incubators
Incubators Compact Benchtop Incubators
IL 10, IL 23 and IL 56 digital incubators offer an economical and space saving solution for microbiology or haematology applications. The temperature can be freely adjusted in increments of 0,1 °C up to a maximum of 70 °C. Heating elements are incorporated into the housing base and walls to ensure temperature stability and uniformity. The IL 10 is available with a transparent Plexiglas® window to provide full visibility or a painted, opaque Plexiglas® window to protect photosensitive samples. IL 23 and IL 56 come as standard with a transparent Plexiglas® window. The housing and door frameMore Product Information