VWR® INCU-Line® IL 23R, Digital Cooled Incubator
Supplier: VWR Collection
VWRI390-0881EA 3820 SGD
VWRI390-0881
VWR® INCU-Line® IL 23R, Digital Cooled Incubator
Incubators Cooled Incubators
The IL 23R digital cooled incubator with Peltier technology offers an economical and space saving solution for microbiology applications at below room temperature. The temperature can be freely adjusted in increments of 0,1 °C from 10 °C below ambient up to a maximum of 50 °C. The built-in Peltier element has a low energy consumption and ensures temperature stability and uniformity over the complete temperature range. The transparent Plexiglas® window allows full visibility of the samples inside the incubator. The housing is constructed entirely of epoxy coated mild steel.