VWR® INCU-Line® ILCO 180 and ILCO260 Premium, CO₂ Incubators
Supplier: VWR Collection
VWRI390-1544EA 17550 SGD
VWRI390-1544
VWR® INCU-Line® ILCO 180 and ILCO260 Premium, CO₂ Incubators
Incubators CO₂ Incubators
The ILCO 180 and ILCO 260 Premium offer optimal growth conditions for cell cultures and meet demanding requirements for best possible cell growth and maximum sample safety. These CO₂ incubators come with a built-in, drift-free infrared CO₂ sensor that can be sterilised. Reliable decontamination is provided by the standard-compliant 180 °C hot air sterilisation routine which completely eliminates contaminants and ensures the inner chamber is completely sterilised. They have an intuitive menu and user friendly software, also USB port for data download to USB stick and for uploading programs.More Product Information