VWR® Cellulose Stoppers
Supplier: VWR Collection
VWRI391-0151EA 101 SGD
VWRI391-0151 VWRI391-0168 VWRI391-0150 VWRI391-0152 VWRI391-0153 VWRI391-0154 VWRI391-0161 VWRI391-0162 VWRI391-0163 VWRI391-0164 VWRI391-0165 VWRI391-0149 VWRI391-0167 VWRI391-0187 VWRI391-0169 VWRI391-0173 VWRI391-0174 VWRI391-0175 VWRI391-0176 VWRI391-0177 VWRI391-0178 VWRI391-0179 VWRI391-0184 VWRI391-0185 VWRI391-0186 VWRI391-0166 VWRI391-0194 VWRI391-0191 VWRI391-0192 VWRI391-0393 VWRI391-0412 VWRI391-0397 VWRI391-0396 VWRI391-0392 VWRI391-0391 VWRI391-0379 VWRI391-0377 VWRI391-0376 VWRI391-0395 VWRI391-0381 VWRI391-0389 VWRI391-0386 VWRI391-0382 VWRI391-0414 VWRI391-0647
VWR® Cellulose Stoppers
Stoppers Cellulose Stoppers
These stoppers are made of cellulose and are used for microbiological samples and tissue cultures in test tubes or Erlenmeyer flasks.
  • Air permeable
  • Autoclavable up to 200 °C
  • Filters bacteria
Ordering information: Available in special or standard shape.
Packaging: Packed in bags and cardboard boxes for easy handling and space saving.
Order Now
SPECIFICATIONS
Environmentally Preferable