VWR®, Petri Dishes, Glass
Supplier: VWR Collection
VWRI391-0576EA 166 SGD
VWRI391-0576 VWRI391-0577 VWRI391-0578 VWRI391-0579 VWRI391-0580 VWRI391-0581
VWR®, Petri Dishes, Glass
Culture Dishes Petri Dishes
Made from borosilicate 3.3 glass that can, therefore, be sterilised and/or used for applications that require heat. These reusable glass Petri dishes are a green alternative to Petri dishes made from plastic, ideal for smaller laboratories.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Autoclavable and sterilisable
Order Now
SPECIFICATIONS
Environmentally Preferable