VWR®, Polyester Films for ELISA and General Incubation
Supplier: VWR Collection
VWRI391-1250EA 146 SGD
VWRI391-1250 VWRI391-1251 VWRI391-0620 VWRI391-0623
VWR®, Polyester Films for ELISA and General Incubation
Seals Microplate Seals
These 77,5 µm thick polyester films provide secure sealing around every well, not just a cover, thus minimising evaporation, preventing spillage and well-to-well contamination, and eliminating 'edge effects' in sensitive ELISA assays. Polyester films can also be used with cell culture plates for short-term storage, incubation, and containment of biohazards.
  • Functional temperature range from −40 to +120 °C
  • Available sterile and non-sterile
  • Non-pierceable
Each film L×D: 146,1×79,4 mm