VWR® Aluminium Foil Seals for PCR
Supplier: VWR Collection
VWRI391-0641EA 1220 SGD
VWRI391-0641
VWR® Aluminium Foil Seals for PCR
Seals Microplate Seals
Pre-cut aluminum foil seals on a roll for PCR applications. Supplied on a roll with holder for ease of use.
  • Ideal for PCR
  • Rapidly seal microplates; faster than applying single sheeted film
  • 2 mil pierceable foil
  • Certified DNase- and RNase-free
  • Recommended temperature range from −20 to +120 °C
Self-standing, non slip dispenser is supplied for ease of use.