VWR® Aluminium Foil Seals for Long-Term Storage, Chemical Resistant (DMSO)
Supplier: VWR Collection
VWRI391-0621EA 283 SGD
VWRI391-0621 VWRI391-0642
VWR® Aluminium Foil Seals for Long-Term Storage, Chemical Resistant (DMSO)
Seals Microplate Seals
Aluminum foils for use with 96-well or 384-well plates. Foils offer excellent chemical resistance (DMSO).
  • Ideal for long-term storage
  • Certified DNase- and RNase-free
  • Temperature range is –80 to +120 °C
  • Pierceable
  • Light blocking
  • No adhesive residue upon removal
391-0621 fits inside the rim of raised rim plates. 391-0642 fits all flat top plates.