VWR® Adhesive Sealing Films for PCR and Storage
Supplier: VWR Collection
VWRI391-0622EA 198 SGD
VWRI391-0622 VWRI391-0643
VWR® Adhesive Sealing Films for PCR and Storage
Seals Microplate Seals
Polyester seals feature a strong adhesive to minimize evaporation during PCR and storage applications.
  • Ideal for PCR & storage
  • Certified DNase- and RNase- free
  • Temperature range is –40 to +120 °C
  • Non-pierceable
  • Minimizes evaporation
Removes easily without leaving adhesive residue.
Order Now
SPECIFICATIONS
Environmentally Preferable