VWR®, Pierceable Films for Robotics, Clear-Zone
Supplier: VWR Collection
VWRI391-1264EA 412 SGD
VWRI391-1264
VWR®, Pierceable Films for Robotics, Clear-Zone
Seals Microplate Seals
With a polyethylene top layer and inert white polypropylene and acrylic adhesive sublayer, this easy piercing sealing film allows direct sample recovery with single- or multi-channel pipettes and robotic probes.
  • Clear zone above each well is free of adhesive
  • Minimise sample exposure to adhesive
  • Recommended temperature range −40 to +90 °C
The adhesive-free zone above each well prevents adhesive fouling of the pipette tip or robotic probe, minimising the amount of adhesive that comes into contact with the sample. Suitable for 96-well plates only.
Order Now
SPECIFICATIONS
Environmentally Preferable