VWR®, Polyester Films with Colour-Coded Marking Areas
Supplier: VWR Collection
VWRI391-1267EA 232 SGD
VWRI391-1267 VWRI391-1268 VWRI391-1269 VWRI391-1270 VWRI391-1271
VWR®, Polyester Films with Colour-Coded Marking Areas
Seals Microplate Seals
Identical to polyester films above, but include a colour dot and white writing area on each end tab for plate identification. End tabs with backing removed adhere to the ends of the plate and are perforated so they can be left in place even if the centre portion of the film is removed. Alternatively, one inscribed colour-coded end tab can be removed and pasted in a laboratory notebook while the other remains on the plate.
  • Choice of five colours for easy plate identification
  • Applications: Short-term storage, incubation and ELISA
  • Recommended temperature range −40...+120 °C