VWR®, Black Film Seals for Fluorescence and Photoprotection
Supplier: VWR Collection
VWRI391-1291EA 176 SGD
VWRI391-1291
VWR®, Black Film Seals for Fluorescence and Photoprotection
Seals Microplate Seals
These films are 240 µm thick black vinyl with adhesive for use with 96-well, 384-well and 1536-well black microplates. They fit within the edges of raised rim plates and may be applied to either the plate top or bottom. The light blocking and absorbing properties of these films make them ideal for reducing stray light and crosstalk in fluorescence applications and for protecting light-sensitive samples in storage.
  • Edge cut outs assist in positioning around ribs on the plate bottom
  • Split backing aids in applying the film properly
  • No end tabs
  • Recommended
More Product Information