VWR®, Beater Mixer Mill
Supplier: VWR Collection
VWRI412-0167EA 11160 SGD
VWRI412-0167
VWR®, Beater Mixer Mill
Homogenisers Homogenisers Bench
A ball mill for disintregration of small sample volumes that can also be used to shake/mix microtubes and microplates. Wide range of applications in different scientific fields from geology to biotechnology.
  • Easy to use - simple knob and keypad to control frequency and timer
  • Easy to maintain - brushless DC motor
  • Large range of grinding jars and tube holders to suit the sample requirements
Grinding time setting - 5 s to 60 min
Vibrational frequency: 3 to 30 Hz