VWR®, Disposable Pestles, Microtubes and Cordless Motor for Pellet Mixing
Supplier: VWR Collection
47747-358
VWRI431-0094EA 296 SGD
VWRI431-0094 VWRI431-0095 VWRI431-0096 VWRI431-0098 VWRI431-0099 VWRU47747-358
VWR®, Disposable Pestles, Microtubes and Cordless Motor for Pellet Mixing
Homogenisers Homogenisers Handheld
Pestles and microtubes manufactured from polybutylene terephthalate (PBTP) in a class 10000, ISO Class 7, M5.5 cleanroom. Designed for homogenising cells and plant tissue or for re-suspending protein and DNA pellets. An optional battery powered motor can be used with the pestles and microtubes. Pestles are 75 mm in length.
  • Free from DNase, RNase and non pyrogenic
  • Grooved grip makes pestle handle easy to turn without slipping, even when wearing gloves
  • Lightweight motor reduces hand fatigue
  • High speed vortexing action of the motor completely homogenises cell
More Product Information