VWR®, Dispersing Aggregates
Supplier: VWR Collection
VWRI431-0280EA 4270 SGD
VWRI431-0280 VWRI431-0278 VWRI431-0279 VWRI431-0281 VWRI431-0282 VWRI431-0283 VWRI431-0284 VWRI431-0285 VWRI431-0286 VWRI431-0287 VWRI431-0288 VWRI431-0289
VWR®, Dispersing Aggregates
Homogenisers Dispersing Tools
Choice of aggregates for a wide range of laboratory applications from 5 ml up to 20 L. The VWR® Aggregates solve the standard problems that are faced every day in the laboratory.
  • Easy to exchange
  • Easy to clean
  • High quality 316L stainless steel
Their universal geometry includes two rows of teeth, so they are suitable for a large spectrum of laboratory applications and are reliable and efficient.
The aggregates with Ø of 5 and 7 mm are for dispersing small amounts, these are suitable for the VWR® Homogeniser S model. Aggregates with Ø of 12 mmMore Product Information