VWR® Homogenisers
Supplier: VWR Collection
VWRI431-0290EA 3400 SGD
VWRI431-0290 VWRI431-0291 VWRI431-0292 VWRI431-0293
VWR® Homogenisers
Homogenisers Homogenisers Bench
These compact homogenisers are suitable for all standard dispersing tasks and intensive mixing. The units are robust, and their stability prevents pulsation oscillations and vibrations, it is also simple and safe to change aggregates for different tasks.
  • Easy operation with an on-off switch and speed control
  • Intelligent variable speed control for long equipment life
  • Low noise level
  • Height adjustable stand (available as an option)
  • Switch aggregate easily
  • Models M, L and XL give speed stability even with changes in viscosity
  • XL model provides protection
More Product Information