VWR®, 4-Place Mini Bead Mill Homogeniser

Supplier: VWR Collection
VWRI432-0366EA 6310 SGD
VWRI432-0366
VWR®, 4-Place Mini Bead Mill Homogeniser
Homogenisers Homogenisers Bench
The VWR® 4-place Mini Bead Mill homogeniser is compact and powerful.

  • Specifically designed for grinding, lysing and the homogenisation of biological samples prior to molecular extraction
  • Ideal solution for releasing DNA, RNA proteins and enzymes from tough samples, while still retaining molecular integrity
  • Offers a broad performance range

Using sample tubes pre-filled with a variety of lysing beads, the mill vigorously and uniformly shakes the tubes providing an efficient, consistent high yield and quality homogenisation in less than 60 seconds. TheMore Product Information
Order Now

SPECIFICATIONS

Model 4-place Mini Bead Mill
Environmentally Preferable
Speed range 1 - 5 m/s
W×D×H 215×292×254 mm
Weight 7,48 kg
Sample volume 25 µl - 5 ml
Run time 1 s - 5 min
Application areas Sample preparation, DNA, RNA, protein extraction
Capacity Max. 4 tubes: 4×0,5 ml / 4×1,5 ml / 4×2 ml / 4×7 ml
Electrical 100 - 230 V, 50/60 Hz

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR