VWR®, Stirring shafts
Supplier: VWR Collection
VWRI441-0253EA 100 SGD
VWRI441-0253 VWRI441-0254 VWRI441-0255 VWRI441-0256 VWRI441-0257 VWRI441-0258 VWRI441-0259 VWRI441-0260 VWRI441-0261
VWR®, Stirring shafts
Stirrers Stirring Shafts
PTFE. For use with stirring blades. Chemically inert. The smooth surface reduces contamination and makes cleaning easier. It also prevents the agitation vessels from being scratched.
  • Excellent chemical resistance
  • Temperature resistant from −200 to 280 °C
Ordering information: Stirring blades must be ordered separately.
Order Now
SPECIFICATIONS
Environmentally Preferable