VWR® VOS 40 digital & VOS 60 control, Electronic Overhead Stirrers
Supplier: VWR Collection
VWRI441-0304EA 2120 SGD
VWRI441-0304 VWRI441-0302 VWRI441-0303 VWRI441-0305 VWRI441-0306 VWRI441-0307
VWR® VOS 40 digital & VOS 60 control, Electronic Overhead Stirrers
Stirrers Overhead Stirrers
Laboratory stirrers that are ideal for simple stirring tasks, suitable for quantities up to 25 L (VOS 40 digital), or for intensive stirring tasks for quantities up to 40 L (VOS 60 control). Due to microprocessor-controlled technology, the stirrers automatically adjust the speed within the speed range of 0/30 to 2000 min⁻¹.
  • Digital speed display
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection - units can be operated for short periods in overload conditions
  • Error code display
Safety circuits ensure automaticMore Product Information