VWR®, Magnetic stirring bars, plain
Supplier: VWR Collection
VWRI442-0262EA 22.6 SGD
VWRI442-0262 VWRI442-0263 VWRI442-0264 VWRI442-0265 VWRI442-0266 VWRI442-0267 VWRI442-0268 VWRI442-0269 VWRI442-0270 VWRI442-0271 VWRI442-0272 VWRI442-0273 VWRI442-0274 VWRI442-0275 VWRI442-0276 VWRI442-0277 VWRI442-0278 VWRI442-0279 VWRI442-0280
VWR®, Magnetic stirring bars, plain
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface with rounded edges that intensify the stirring effect, even at low speeds.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Order Now
SPECIFICATIONS
Environmentally Preferable