VWR®, Magnetic stirring bars, cylindrical, with removable pivot ring
Supplier: VWR Collection
VWRI442-0375EA 108 SGD
VWRI442-0375 VWRI442-0369 VWRI442-0371 VWRI442-0373 VWRI442-0374 VWRI442-0376 VWRI442-0378 VWRI442-0379 VWRI442-0381 VWRI442-0382 VWRI442-0383 VWRI442-0384
VWR®, Magnetic stirring bars, cylindrical, with removable pivot ring
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface. For containers with slightly uneven bases.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Order Now
SPECIFICATIONS
Environmentally Preferable