VWR®, Magnetic stirring bars, triangular
Supplier: VWR Collection
VWRI442-0385EA 36.1 SGD
VWRI442-0385 VWRI442-0386 VWRI442-0387 VWRI442-0388 VWRI442-0389 VWRI442-0390 VWRI442-0391 VWRI442-0392 VWRI442-0393
VWR®, Magnetic stirring bars, triangular
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface. Particularly effective for dissolving solids and mixing sediments because of the scraper-like action on the base of the container.
  • High turbulence, even at low speeds
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Order Now
SPECIFICATIONS
Environmentally Preferable