VWR®, Magnetic Stirring Bars, Cross-Shaped
Supplier: VWR Collection
VWRI442-0394EA 36.2 SGD
VWRI442-0394 VWRI442-0395 VWRI442-0396 VWRI442-0397 VWRI442-0398 VWRI442-0399 VWRI442-0400
VWR®, Magnetic Stirring Bars, Cross-Shaped
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Smooth surface. Stable rotating position for optimal stirring effect.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Order Now
SPECIFICATIONS
Environmentally Preferable