VWR®, Magnetic stirring bars, oval
Supplier: VWR Collection
VWRI442-0401EA 24.7 SGD
VWRI442-0401 VWRI442-0402 VWRI442-0403 VWRI442-0404 VWRI442-0405 VWRI442-0406 VWRI442-0407 VWRI442-0408 VWRI442-0409 VWRI442-0410
VWR®, Magnetic stirring bars, oval
Stirrers Magnetic Stirring Bars
PTFE covered, Alnico V magnetic core. Smooth surface. Designed for use in round bottomed flasks.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Order Now
SPECIFICATIONS
Environmentally Preferable