VWR®, Magnetic stirring discs
Supplier: VWR Collection
VWRI442-0490EA 81.8 SGD
VWRI442-0490 VWRI442-0491 VWRI442-0492
VWR®, Magnetic stirring discs
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. The special shape with raised surface in the centre ensures very good agitation even at low speeds. Ideal for use in glass beakers.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Order Now
SPECIFICATIONS
Environmentally Preferable