VWR®, Magnetic Wing Stirrers, Micro
Supplier: VWR Collection
VWRI442-0493EA 68.4 SGD
VWRI442-0493 VWRI442-0494 VWRI442-0495 VWRI442-0496 VWRI442-0497
VWR®, Magnetic Wing Stirrers, Micro
Stirrers Magnetic Stirring Bars
PTFE covered, Alnico V magnetic core. Smooth surface. For stirring in test tubes, centrifuge tubes and micro vials. The wings can also be trimmed to size if needed.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)