VWR®, Magnetic stirrer for cuvettes
Supplier: VWR Collection
VWRI442-0502EA 34.9 SGD
VWRI442-0502
VWR®, Magnetic stirrer for cuvettes
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Designed for agitating in 10 mm standard cuvettes.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)