VWR®, Magnetic Stirring Bars, Elliptical, Rare Earth
Supplier: VWR Collection
VWRI442-0503EA 58.8 SGD
VWRI442-0503 VWRI442-0504 VWRI442-0505 VWRI442-0506 VWRI442-0507
VWR®, Magnetic Stirring Bars, Elliptical, Rare Earth
Stirrers Magnetic Stirring Bars
PTFE covered, rare earth magnetic core. The magnetic core is made from a combination of samarium and cobalt. The stirrers will continue to function at full strength for many years as the magnetism remains for longer. They are identified by an inert carbon black spot.
  • Magnetic attraction is 1,5 to 2 times stronger than conventional stirrers
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Order Now
SPECIFICATIONS
Environmentally Preferable