VWR®, Magnetic stirring bars, glass-covered
Supplier: VWR Collection
VWRI442-0508EA 118 SGD
VWRI442-0508 VWRI442-0509 VWRI442-0510 VWRI442-0511 VWRI442-0512
VWR®, Magnetic stirring bars, glass-covered
Stirrers Magnetic Stirring Bars
Cylindrical, with Alnico V magnetic core.
  • Poreless, track neutral, extremely smooth surface
  • Excellent chemical resistance
Order Now
SPECIFICATIONS
Environmentally Preferable