VWR®, Magnetic stirring bars, micro
Supplier: VWR Collection
VWRI442-0359EA 30.7 SGD
VWRI442-0359 VWRI442-0360 VWRI442-0361 VWRI442-0362 VWRI442-0363 VWRI442-0364 VWRI442-0365 VWRI442-0366 VWRI442-0367 VWRI442-0368 VWRI442-0447 VWRI442-0448 VWRI442-0449 VWRI442-0450 VWRI442-0451 VWRI442-0452 VWRI442-0453 VWRI442-0454 VWRI442-0455 VWRI442-0456 VWRI442-0457 VWRI442-0458 VWRI442-0459 VWRI442-0460 VWRI442-0461 VWRI442-0462 VWRI442-0463 VWRI442-0464 VWRI442-0465 VWRI442-0466 VWRI442-0467 VWRI442-0468 VWRI442-0469 VWRI442-0470 VWRI442-0471 VWRI442-0472 VWRI442-0473 VWRI442-0594 VWRI442-0595 VWRI442-0596 VWRI442-0597
VWR®, Magnetic stirring bars, micro
Stirrers Magnetic Stirring Bars
PTFE coated, strong Alnico V magnetic core. Round, smooth surface. For use in the smallest containers.
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Order Now
SPECIFICATIONS
Environmentally Preferable