VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
VWRI442-0661EA 1900 SGD
VWRI442-0661 VWRI442-0662 VWRI442-0663 VWRI442-0664 VWRI442-0665 VWRI442-0666 VWRI442-0667 VWRI442-0668 VWRI442-0669
VWR® VMS Advanced Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
Compact stirrers with a ceramic glass plate provide excellent chemical resistance. Units feature digital error code display, a 'hot' top indicator to warn users that surface is hot, a fixed safety circuit that shuts off the heater if temperature rises above 550 °C, and three operating modes to choose from (standard, safe and adjustment protection). Stirrers have a speed display scale and optional capability to connect directly to a PT1000 temperature sensor; when using this functionality, control accuracy in the medium is ±0,5 °C.
  • Exact temperature setting via digital potentiometer
More Product Information