VWR®, Advanced hotplates
Supplier: VWR Collection
VWRI442-1269EA 1460 SGD
VWRI442-1269
VWR®, Advanced hotplates
Heaters Hotplates
VWR® Advanced hotplates are designed to deliver accurate and repeatable results in all research, academic and industrial applications. With temperature ranges up to 500 °C VWR hotplates provide superior temperature control.
  • Excellent temperature uniformity
  • Easy to read, large LCD displays temperatures
  • Industry-leading safety features
  • Cool touch and chemical resistant housing
  • 5 year warranty
These microprocessor-controlled units feature a front facing control panel with an easy to read LCD that displays temperature settings and intuitive icons.