VWR®, Advanced Magnetic Stirrers
Supplier: VWR Collection
VWRI442-1310EA 1590 SGD
VWRI442-1310
VWR®, Advanced Magnetic Stirrers
Stirrers Magnetic Stirrers
VWR® Advanced magnetic stirrers are designed to deliver accurate and repeatable results in all research, academic and industrial applications. With stirring speeds reaching 1600 rpm, VWR® magnetic stirrers provide proper mixing. Available with resin plate.
  • Excellent stirring
  • Easy to read, large LCD displays speed
  • Cool touch and chemical resistant housing
These microprocessor-controlled units feature a front facing control panel with an easy to read LCD that displays speed settings and intuitive icons.
Magnetic stirrers are available with resin topMore Product Information