VWR®, Magnetic stirring bars, cylindrical
Supplier: VWR Collection
VWRI442-4520EA 21.2 SGD
VWRI442-4520 VWRI442-4521 VWRI442-4522 VWRI442-4523 VWRI442-4524 VWRI442-4525 VWRI442-4530 VWRI442-4527 VWRI442-4528 VWRI442-4529 VWRI442-0259 VWRI442-0260 VWRI442-0261 VWRI442-0295
VWR®, Magnetic stirring bars, cylindrical
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Rounded, smooth surface.
  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Order Now
SPECIFICATIONS
Environmentally Preferable