VWR® lab dancer, Vortex Mixer
Supplier: VWR Collection
VWRI444-0004EA 643 SGD
VWRI444-0004
VWR® lab dancer, Vortex Mixer
Shakers Vortex Shakers
Vortex mixer suitable for single, small containers up to 30 mm in diameter, for example, test tubes, centrifuge tubes and microtubes. The upper casing is PP and the test tube surface is made from inert TPU-plastic; the bottom section is a coated, zinc die-casting.
  • Small, compact and reliable
  • Excellent mixing action
IP protection class according to DIN EN 60529: IP 40
Certifications: IP protection class according to DIN EN 60529: IP 40
Delivery information: Supplied with 12 V power pack set.