VWR® Incubating 3-D Rotator Waver
Supplier: VWR Collection
VWRI444-0765EA 5480 SGD
VWRI444-0765 VWRI444-0766 VWRI444-0767
VWR® Incubating 3-D Rotator Waver
Shakers Shaking Incubators
The incubating three-dimensional rotator waver combines vertical and horizontal wave motion with incubation in one compact bench top unit. The spill resistant design channels fluids away from internal components. The microprocessor control provides electronic tilt angle adjustment which allows the user to easily adjust rocking angle from 0 to 20° while unit is operating. The PID temperature controller maintains precise temperature control from ambient 5 to 65 °C.
  • Electronic tilt adjustment from 0 to 20° while unit is operating
  • LED displays shows
More Product Information