VWR®, Support Stand, Rectangular Base, Cast Iron
Supplier: VWR Collection
VWRI444-0918EA 423 SGD
VWRI444-0918
VWR®, Support Stand, Rectangular Base, Cast Iron
Retort Stands
Cast iron support stand bases are constructed with black enamel finish for chemical resistance and durability.
It features a built-in support rod holder with locking knob that accepts a 13 mm diameter support rod.
Ordering information: Support rods are not included.