VWR®, Support Rods
Supplier: VWR Collection
VWRI444-0919EA 173 SGD
VWRI444-0919
VWR®, Support Rods
Retort Rods
Heavy-duty, top-of-the-line rods provide maximum strength and durability. Constructed of 303 stainless steel.