VWR®, Mini vortex mixer
Supplier: VWR Collection
VWRI444-1337EA 348 SGD
VWRI444-1337
VWR®, Mini vortex mixer
Shakers Vortex Shakers
The mini vortex mixer is the ideal vortex mixer for every laboratory with its compact size, ease of use, and its economic price point. The 3000 min⁻¹ fixed speed ensures there is no sacrificing any mixing power.
  • Extremely compact and easy to operate
  • Powerful vortexing for tubes up to Ø 30 mm
  • Refined steel base is designed to ensure product stability