VWR® Test Tube Racks, Pivoting Stainless Steel
Supplier: VWR Collection
VWRI444-2955EA 849 SGD
VWRI444-2955 VWRI444-2956 VWRI444-2957 VWRI444-2958 VWRI444-2959
VWR® Test Tube Racks, Pivoting Stainless Steel
Racks Tube Racks
Accessories information: * Platform supplied with mini shaker, incubating mini shaker, microplate shaker and incubating microplate shaker. Note other shakers require an optional perforated platform to enable flask clamps or test tube racks to be mounted which must be ordered separately.