VWR® Stainless Steel Erlenmeyer Flask Clamps
Supplier: VWR Collection
57018-822
VWRI444-7040EA 91.6 SGD
VWRI444-7040 VWRI444-7041 VWRI444-7042 VWRI444-7043 VWRI444-7044 VWRI444-7045 VWRU57018-822
VWR® Stainless Steel Erlenmeyer Flask Clamps
Clamps General Purpose Clamps
Accessories information: * Platform supplied with mini shaker and incubating mini shaker. Note other shakers require an optional universal platform to enable flask clamps or test tube racks to be mounted which must be ordered separately.
Order Now
SPECIFICATIONS
Environmentally Preferable
Standard platform 330×330 mm, capacity 60 30 15 12 8