VWR®, Orbital Mini Shaker
Supplier: VWR Collection
VWRI444-0268EA 2190 SGD
VWRI444-0268 VWRI444-0269 VWRI444-7093
VWR®, Orbital Mini Shaker
Shakers Orbital Shakers
General purpose shaker with microprocessor control which is ideal for a range of mixing applications. Variable speed control provides consistent, uniform shaking action. The speed is adjustable in 10 min⁻¹ increments up to 500 min⁻¹, and increments of 25 min⁻¹ up to 1200 min⁻¹. The ramping feature slowly increases speed to the set point for added safety.
  • Two independent LED displays show speed and time
  • Integral timer, 1 s to 160 h, shows elapsed time or counts down and enables automatic switch off and audible alarm when program is
More Product Information