VWR® Advanced Mini Shaker 15, Orbital Shaker
Supplier: VWR Collection
VWRI444-7106EA 3640 SGD
VWRI444-7106 VWRI444-7107
VWR® Advanced Mini Shaker 15, Orbital Shaker
Shakers Orbital Shakers
A compact, powerful mini shaker with microprocessor control that is ideal for a wide variety of shaking applications. The unit has optimal speed range and orbit for larger samples. Variable speed control provides consistent, uniform shaking action. The speed is adjustable in 1 min⁻¹ increments throughout the speed range. Ramping feature slowly increases speed to desired set point to avoid splashing.
  • 15 mm orbit gives optimal speed for mixing larger-sized samples, unit provides powerful shaking with a small footprint
  • Independent LED displays for speed, time,
More Product Information