VWR® VHP-C, Hotplates
Supplier: VWR Collection
VWRI453-0050EA 1450 SGD
VWRI453-0050 VWRI453-0051 VWRI453-0053 VWRI453-0054 VWRI453-0056 VWRI453-0057
VWR® VHP-C, Hotplates
Heaters Hotplates
Compact hotplates with a chemically resistant ceramic top. The elevated control panel minimises contact with spills and enables easy reading and adjustment of the temperature. Units feature a digital error code display and 'hot' top indicator to warn users that the surface is hot.
  • VHP-C7 can be used with the VT-5 contact thermometer (442-0573), enabling precise temperature control
  • Fixed safety circuit shuts off heater if temperature rises above 550 °C
  • Microprocessor control sends feedback to the hotplate to maintain set temperature
  • Digital display ensures
More Product Information