VWR®, Crucibles, Porcelain
Supplier: VWR Collection
VWRI459-0209EA 39.8 SGD
VWRI459-0209 VWRI459-0208 VWRI459-0207 VWRI459-0198 VWRI459-0197 VWRI459-0196 VWRI459-0195 VWRI459-0194 VWRI459-0201 VWRI459-0210 VWRI459-0214 VWRI459-0213 VWRI459-0199 VWRI459-0211 VWRI459-0200 VWRI459-0206 VWRI459-0205 VWRI459-0204 VWRI459-0203 VWRI459-0202 VWRI459-0212
VWR®, Crucibles, Porcelain
Crucibles
Porcelain, glazed inside and out, except base.
  • Withstands maximum temperature of 1000 °C
Lids are available separately.
Lids are available separately.
Certifications: According to DIN 12904.
Order Now
SPECIFICATIONS
Environmentally Preferable