VWR®, Crucibles, Stainless Steel 304
Supplier: VWR Collection
VWRI459-4501EA 33 SGD
VWRI459-4501 VWRI459-4502 VWRI459-4503
VWR®, Crucibles, Stainless Steel 304
Crucibles
These crucibles are made from stainless steel 304.
  • Highly resistant to chemicals and reagents
  • Withstand maximum temperature of 800 °C
Without lid.