VWR®, Block Heaters, Digital
Supplier: VWR Collection
VWRI460-0349EA 1670 SGD
VWRI460-0349 VWRI460-0350 VWRI460-0351 VWRI460-0352
VWR®, Block Heaters, Digital
Block Heaters Digital Block Heaters
Multi-purpose, high wattage, constant temperature, VWR® digital block heaters are economical, versatile and compact. These digital models provide reproducable results, exceptional temperature uniformity, and are ideal for a wide range of applications. Temperature calibration mode allows the user to calibrate the unit to an external temperature device. In timed mode, an alarm will sound when the time reaches zero and the unit reaches set point temperature.
  • Compact design with small footprint
  • Temperature range from ambient +5 to 120 °C
  • 'Hot' symbol warning
More Product Information