VWR®, Dry Block Heater, Digital, with Heated Lid
Supplier: VWR Collection
VWRI460-0353EA 4140 SGD
VWRI460-0353
VWR®, Dry Block Heater, Digital, with Heated Lid
Block Heaters Digital Block Heaters
Multi-purpose, high wattage, constant temperature, VWR® digital block heater which is economical, versatile and compact. This digital model, with heated lid, provides reproducable results, exceptional temperature uniformity and is ideal for a wide range of applications. Temperature calibration mode allows the user to calibrate the unit to an external temperature device. In timed mode, an alarm will sound when the time reaches zero and the unit reaches the set point temperature.
  • Compact design with small footprint
  • Temperature range from ambient 5 to 100 °C
  • '
More Product Information